Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167939.1 | NP_001161411.1 | 7 | 745 | ||
NM_028993.4 | NP_083269.4 | 7 | 745 | |||
GenBank mRNA | AK018662.1 | - | 7 | 713 | ||
AK034837.1 | - | 7 | 707 | |||
AK039196.1 | - | 7 | 711 | |||
AK049277.1 | - | 7 | 706 | |||
AK081967.1 | - | 7 | 746 | |||
AK162318.1 | - | 6 | 1145 | |||
AK164005.1 | - | 7 | 712 | |||
BC125568.1 | - | 7 | 745 | 40130083 | ||
BC132139.1 | - | 7 | 745 | 40130416 | ||
BC144990.1 | - | 7 | 745 | 9053538 |