Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164366.1 | NP_001157838.1 | 5, 6 | 663 | ||
NM_029097.2 | NP_083373.2 | 5, 6 | 663 | |||
GenBank mRNA | AK152421.1 | - | 16, 17 | 1871 | ||
AK156104.1 | - | 5, 6 | 643 | |||
AK219794.1 | - | 1, 2 | 85 | |||
BC023746.1 | - | 5, 6 | 638 | 5353766 | ||
BC025028.1 | - | 5, 6 | 558 | |||
BC042661.1 | - | 5, 6 | 608 | 4192094 |