Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029238.2 | NP_083514.1 | 6 | 1196 | ||
GenBank mRNA | AB041538.1 | - | 7 | 1124 | ||
AK016089.1 | - | 3 | 390 | |||
AK034308.1 | - | 6 | 1198 | |||
BC138981.1 | - | 6 | 1095 | 8862003 | ||
BC145720.1 | - | 6 | 1095 | 40131023 |