Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029364.3 | NP_083640.1 | 9 | 1109 | ||
GenBank mRNA | AK030773.1 | - | 9 | 1108 | ||
AK049162.1 | - | 9 | 1110 | |||
AK054046.1 | - | 9 | 1109 | |||
AK083597.1 | - | 9 | 1087 | |||
AK149863.1 | - | 9 | 1094 | |||
AK151037.1 | - | 7 | 731 | |||
AK159562.1 | - | 9 | 1076 | |||
AK165180.1 | - | 9 | 1089 | |||
AK165917.1 | - | 9 | 1059 | |||
AK169485.1 | - | 9 | 1091 | |||
AK170791.1 | - | 9 | 1105 | |||
AK197098.1 | - | 2 | 77 | |||
AK198818.1 | - | 2 | 77 | |||
AK208382.1 | - | 2 | 77 | |||
BC055328.1 | - | 9 | 1103 | 6400020 | ||
BC059047.1 | - | 1 | 263 |