Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175224.4 | NP_780433.1 | 2 | 155 | ||
GenBank mRNA | AK077694.1 | - | 2 | 156 | ||
AK152138.1 | - | 2 | 174 | |||
AK153231.1 | - | 2 | 174 | |||
AK153944.1 | - | 2 | 163 | |||
AK165034.1 | - | 2 | 172 | |||
BC096469.1 | - | 2 | 146 | 3603051 |