Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081241.2 | NP_001074710.2 | 3 | 377 | ||
GenBank mRNA | AK052316.1 | - | 3 | 326 | ||
AK122573.1 | - | 3 | 351 | |||
BB852730.1 | - | 3 | 376 | |||
BC079880.1 | - | 3 | 291 | 6852251 |