Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113423.2 | NP_001106894.1 | 7 | 1691 | ||
NM_153567.3 | NP_705795.2 | 8 | 1769 | |||
GenBank mRNA | AK129365.1 | - | 7 | 1680 | ||
AK146432.1 | - | 7 | 1690 | |||
BC019454.1 | - | 1 | 108 | |||
BC038019.1 | - | 6 | 974 | 3597423 | ||
BC076573.1 | - | 7 | 1624 | |||
CK619668.1 | - | 4 | 476 |