Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199217.1 | NP_001186146.1 | 14 | 1391 | ||
NM_001199218.1 | NP_001186147.1 | 14 | 1411 | |||
NM_023805.3 | NP_076294.2 | 14 | 1559 | |||
GenBank mRNA | AF159856.1 | - | 14 | 1361 | ||
AK002607.1 | - | 14 | 1377 | |||
AK035155.1 | - | 1579 | ||||
BC010678.1 | - | 14 | 1390 | |||
BC021324.1 | - | 11 | 2075 | |||
BC054846.1 | - | 14 | 1534 | 6493334 | ||
BC055339.1 | - | 14 | 1411 | 6415115 |