Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029570.3 | NP_083846.2 | 19 | 2465 | ||
GenBank mRNA | AK043943.1 | - | 8 | 1113 | ||
AK129248.1 | - | 19 | 2457 | |||
AK154156.1 | - | 19 | 2471 | |||
AK155372.1 | - | 19 | 2469 | |||
BC026986.1 | - | 4 | 534 | |||
BC076603.1 | - | 19 | 2335 | 30629594 |