Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029620.2 | NP_083896.1 | 3 | 372 | ||
GenBank mRNA | AF352788.1 | - | 3 | 268 | ||
AK010249.1 | - | 3 | 373 | |||
AK132395.1 | - | 262 | ||||
BC051174.1 | - | 3 | 245 | 6310294 |