Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt8d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165930.1 | NP_001159402.1 | 10 | 1102 | ||
NM_029626.2 | NP_083902.2 | 10 | 1631 | |||
GenBank mRNA | AK010388.1 | - | 10 | 1360 | ||
AK077388.1 | - | 10 | 1095 | |||
BC029215.1 | - | 8 | 1303 | |||
BC055709.1 | - | 9 | 1518 | |||
BC058731.1 | - | 9 | 1518 | |||
BC069873.1 | - | 10 | 1181 | 6314675 |