Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081960.1 | NP_001075429.1 | 33 | 3910 | ||
NM_001114347.1 | NP_001107819.1 | 39 | 4652 | |||
NM_001286600.1 | NP_001273529.1 | 34 | 3961 | |||
NM_001286601.1 | NP_001273530.1 | 34 | 3927 | |||
NM_001286602.1 | NP_001273531.1 | 34 | 3973 | |||
NM_029633.2 | NP_083909.2 | 32 | 3907 | |||
GenBank mRNA | AB093252.1 | - | 30 | 3496 | ||
AJ276961.1 | - | 15 | 1910 | |||
AK043551.1 | - | 32 | 3907 | |||
BC026688.1 | - | 4 | 538 | |||
BC117963.1 | - | 33 | 3898 | 40098296 | ||
BC117964.1 | - | 34 | 3949 | 40098298 | ||
BC131791.1 | - | 33 | 3898 | 40098299 | ||
BC131792.1 | - | 34 | 3915 | 40098301 | ||
BC141415.1 | - | 34 | 3959 | 9056028 |