Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029639.2 | NP_083915.2 | 4 | 819 | ||
GenBank mRNA | AK005558.1 | - | 4 | 822 | ||
AY364436.1 | - | 4 | 819 | |||
BC017624.1 | - | 4 | 803 | 4487871 | ||
BC022950.1 | - | 4 | 799 | 5148408 |