Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175251.4 | NP_780460.3 | 15 | 3680 | ||
GenBank mRNA | AK014481.1 | - | 2 | 1645 | ||
AK077366.1 | - | 1 | 442 | |||
AK173201.1 | - | 1 | 1507 | |||
BC027180.1 | - | 1 | 579 | 4021271 |