Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029961.2 | NP_084237.1 | 4 | 278 | ||
GenBank mRNA | AK020318.1 | - | 4 | 279 | ||
AY766239.1 | - | 4 | 242 |