Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030026.2 | NP_084302.1 | 4, 5 | 518 | ||
GenBank mRNA | AK132265.1 | - | 4, 5 | 498 | ||
AK143233.1 | - | 4, 5 | 519 | |||
AK145564.1 | - | 4, 5 | 470 | |||
AK146844.1 | - | 4, 5 | 478 | |||
AK146865.1 | - | 4, 5 | 468 | |||
BC027640.1 | - | 4, 5 | 453 | |||
BC141193.1 | - | 4, 5 | 484 | 9055805 | ||
BC141194.1 | - | 4, 5 | 484 | 9055806 | ||
BY713615.1 | - | 4, 5 | 498 | |||
CK620826.1 | - | 4, 5 | 473 |