Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207204.2 | NP_997087.2 | 14, 15 | 1996 | ||
GenBank mRNA | BC067071.1 | - | 14, 15 | 1930 | 30615484 |