Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142728.1 | NP_001136200.1 | 19 | 2721 | ||
NM_030070.3 | NP_084346.2 | 19 | 2724 | |||
GenBank mRNA | AK019545.1 | - | 19 | 2657 | ||
AK029802.1 | - | 19 | 2722 |