Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082476.2 | NP_001075945.1 | 8 | 1047 | ||
NM_001252541.1 | NP_001239470.1 | 6 | 846 | |||
NM_001252542.1 | NP_001239471.1 | 8 | 1114 | |||
GenBank mRNA | AK019617.1 | - | 8 | 1044 | ||
AK143124.1 | - | 8 | 1088 | |||
AK163207.1 | - | 6 | 816 | |||
AK169885.1 | - | 8 | 1106 | |||
BC049789.1 | - | 8 | 1097 | 6331033 |