Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023912.2 | NP_076401.1 | 10 | 1618 | ||
GenBank mRNA | AF276514.1 | - | 10 | 1618 | ||
AK019240.1 | - | 10 | 1611 | |||
AK134356.1 | - | 10 | 1617 | |||
AK144134.1 | - | 10 | 1604 | |||
AK145869.1 | - | 10 | 1638 | |||
AK161813.1 | - | 10 | 1617 | |||
AK177237.1 | - | 1 | 91 | |||
BC034519.1 | - | 10 | 1604 | 3672524 | ||
HM856325.1 | - | 10 | 1618 | |||
HM856326.1 | - | 10 | 1618 |