Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080964.1 | NP_001074433.1 | 4 | 1197 | ||
NM_030220.3 | NP_084496.2 | 4 | 1200 | |||
GenBank mRNA | AK019649.2 | - | 5 | 1413 | ||
AK078246.1 | - | 4 | 1180 | |||
AK080877.1 | - | 4 | 1208 | |||
AK139302.1 | - | 4 | 1167 | |||
AK156580.1 | - | 4 | 1182 | |||
AK172877.1 | - | 4 | 1171 | |||
BC021759.1 | - | 4 | 1167 | 5151210 | ||
BC086457.1 | - | 4 | 1166 | 6490946 | ||
GU126673.1 | - | 4 | 1156 |