Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nadsyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ305343.1 | - | 2 | 225 | ||
AK020264.1 | - | 2 | 225 | |||
AK041101.1 | - | 2 | 212 | |||
AK042389.1 | - | 2 | 185 | |||
AK045774.1 | - | 2 | 212 | |||
AK046491.1 | - | 2 | 212 | |||
BC038016.1 | - | 2 | 171 | 3589586 | ||
BY035654.1 | - | 2 | 235 |