Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030225.4 | NP_084501.1 | 15 | 1365 | ||
GenBank mRNA | AK019713.2 | - | 1362 | |||
AK054053.1 | - | 15 | 1319 | |||
AK149664.1 | - | 15 | 1341 | |||
AK158877.1 | - | 15 | 1366 | |||
AK168570.1 | - | 15 | 1341 | |||
AK169943.1 | - | 15 | 1319 | |||
AK193525.1 | - | 1 | 26 | |||
AK212893.1 | - | 1 | 26 | |||
AK218237.1 | - | 1 | 26 | |||
BC006702.1 | - | 15 | 1302 | 3583936 | ||
BC024066.1 | - | 6 | 752 | |||
CT010197.1 | - | 15 | 1301 |