Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175283.3 | NP_780492.2 | 1 | 264 | ||
GenBank mRNA | AK082819.1 | - | 1 | 156 | ||
BC079863.1 | - | 1 | 242 | 6807475 | ||
BC094503.1 | - | 243 | 5053446 | |||
BC115468.1 | - | 1 | 121 | 40100423 | ||
BC115469.1 | - | 1 | 121 | 40100424 |