Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckipsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030729.4 | NP_109654.2 | 3 | 529 | ||
GenBank mRNA | AF130313.2 | - | 3 | 425 | ||
AK134725.1 | - | 3 | 511 | |||
BC064818.1 | - | 3 | 529 | 30292108 |