Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030732.3 | NP_109657.2 | 11 | 1387 | ||
GenBank mRNA | AF268195.1 | - | 11 | 1168 | ||
AK029595.1 | - | 11 | 1186 | |||
AK033347.1 | - | 11 | 1257 | |||
AK036064.1 | - | 12 | 1498 | |||
AK136255.1 | - | 11 | 1209 | |||
BC018512.1 | - | 1 | 33 |