Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148930.3 | NP_683732.1 | 19, 20 | 1936 | ||
GenBank mRNA | AJ309168.1 | - | 18, 19 | 1771 | ||
AK134183.1 | - | 19, 20 | 1931 | |||
AK146880.1 | - | 19, 20 | 1931 | |||
BC003988.1 | - | 9, 10 | 885 | |||
BC005719.1 | - | 14, 15 | 1541 | |||
BC023854.1 | - | 19, 20 | 1921 | 5346780 | ||
BC031899.1 | - | 19, 20 | 1921 | 5362121 | ||
BC042419.1 | - | 17, 18 | 5172 | |||
BC043058.1 | - | 19, 20 | 1873 | 6419695 | ||
BC054729.1 | - | 19, 20 | 1924 | 6419758 | ||
BC054820.1 | - | 17, 18 | 4650 | |||
BC060985.1 | - | 18, 19 | 1840 |