Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109909.1 | NP_001103379.1 | 7, 8 | 1221 | ||
NM_001109910.1 | NP_001103380.1 | 7, 8 | 1221 | |||
NM_031405.2 | NP_113582.1 | 7, 8 | 1221 | |||
GenBank mRNA | AK144300.1 | - | 7, 8 | 1212 | ||
BC066831.1 | - | 7, 8 | 1180 | 30471339 |