Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145800.1 | NP_001139272.1 | 5 | 674 | ||
NM_033608.3 | NP_291086.2 | 5 | 740 | |||
GenBank mRNA | AF317839.1 | - | 5 | 740 | ||
AK053544.1 | - | 4 | 1291 | |||
AK078536.1 | - | 5 | 668 | |||
AK145121.1 | - | 5 | 649 | |||
BC034594.1 | - | 5 | 656 | |||
BC040281.1 | - | 5 | 700 | 5364073 |