Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053142.3 | NP_444372.2 | 1 | 755 | ||
GenBank mRNA | AF326310.1 | - | 1 | 617 | ||
AK077255.1 | - | 1 | 726 | |||
AK122533.1 | - | 981 | ||||
AY013778.1 | - | 1 | 617 | |||
BC017149.1 | - | 1 | 765 | 4506788 |