Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053147.3 | NP_444377.3 | 1 | 739 | ||
GenBank mRNA | AK043492.1 | - | 1 | 740 | ||
AK143176.1 | - | 1 | 1220 | |||
AY013784.1 | - | 1 | 611 | |||
BC094239.1 | - | 2 | 212 | 30606144 |