Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110146.1 | NP_001103616.1 | 11 | 1146 | ||
NM_021400.3 | NP_067375.3 | 12 | 3540 | |||
GenBank mRNA | AB034730.1 | - | 13 | 3039 | ||
AK132597.1 | - | 6 | 1286 | |||
AK149469.1 | - | 11 | 1147 | |||
BC130021.1 | - | 10 | 1110 |