Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133787.2 | NP_598548.1 | 16 | 1536 | ||
GenBank mRNA | AK034772.1 | - | 736 | |||
AK088667.1 | - | 16 | 1549 | |||
AK145248.1 | - | 16 | 1558 | |||
AK166283.1 | - | 16 | 1504 | |||
BC003842.1 | - | 16 | 1536 | 3495021 |