Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133807.1 | NP_598568.1 | 3 | 436 | ||
GenBank mRNA | AK053450.1 | - | 3 | 463 | ||
AK159374.1 | - | 3 | 492 | |||
AK159552.1 | - | 3 | 486 | |||
AK166576.1 | - | 3 | 485 | |||
AK167604.1 | - | 3 | 427 | |||
AK169250.1 | - | 3 | 411 | |||
AK169390.1 | - | 3 | 435 | |||
BC006877.1 | - | 3 | 436 | 2647513 | ||
BC066172.1 | - | 3 | 430 | 30429192 |