Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290683.1 | NP_001277612.1 | 3 | 264 | ||
NM_028534.3 | NP_082810.1 | 3 | 534 | |||
GenBank mRNA | AK014888.1 | - | 3 | 218 | ||
AK014966.1 | - | 3 | 218 | |||
AK132868.1 | - | 3 | 265 | |||
BC006946.1 | - | 3 | 442 | 2810817 | ||
BC100537.1 | - | 470 |