Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiparp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178892.5 | NP_849223.2 | 6 | 2002 | ||
GenBank mRNA | AK028529.1 | - | 6 | 2003 | ||
AK032453.1 | - | 6 | 1986 | |||
AK162094.1 | - | 6 | 2008 | |||
AK163347.1 | - | 6 | 2041 | |||
AK186495.1 | - | 2 | 363 | |||
BC068173.1 | - | 6 | 1976 | 30616032 |