Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153597.2 | NP_705825.2 | 4 | 782 | ||
GenBank mRNA | AK038830.1 | - | 3 | 614 | ||
AK048558.1 | - | 931 | ||||
AK090183.1 | - | 3 | 1913 | |||
AK141582.1 | - | 569 | ||||
BC027575.1 | - | 4 | 580 | 1364225 |