Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252374.1 | NP_001239303.1 | 5 | 294 | ||
NM_026004.3 | NP_080280.3 | 4 | 451 | |||
GenBank mRNA | AK011525.1 | - | 4 | 402 | ||
AK011894.1 | - | 4 | 349 | |||
AK160139.1 | - | 5 | 294 | |||
AK216181.1 | - | 1 | 30 | |||
BC038029.1 | - | 5 | 287 | 4018956 | ||
CB571852.1 | - | 5 | 299 | |||
CX734167.1 | - | 5 | 401 |