Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025898.3 | NP_080174.1 | 4, 5 | 369 | ||
GenBank mRNA | AK004826.1 | - | 4, 5 | 378 | ||
AK005372.1 | - | 4, 5 | 371 | |||
AK013895.1 | - | 4, 5 | 388 | |||
AK050623.1 | - | 4, 5 | 433 | |||
AK136689.1 | - | 4, 5 | 372 | |||
AK171998.1 | - | 4, 5 | 433 | |||
BC004804.1 | - | 4, 5 | 400 | 3584869 |