Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113246.2 | NP_001106717.2 | 13 | 1571 | ||
NM_001166603.1 | NP_001160075.1 | 12 | 1397 | |||
NM_001166604.1 | NP_001160076.1 | 6 | 1209 | |||
NM_029716.3 | NP_083992.1 | 6 | 772 | |||
NM_175752.3 | NP_786928.2 | 7 | 1383 | |||
GenBank mRNA | AB264771.1 | - | 6 | 1033 | ||
AB264772.1 | - | 12 | 1094 | |||
AB264773.1 | - | 3740 | ||||
AB264774.1 | - | 5 | 1173 | |||
AF332069.1 | - | 7 | 891 | |||
AF332070.1 | - | 7 | 891 | |||
AK007182.1 | - | 6 | 772 | |||
AK032630.1 | - | 7 | 1341 | |||
AK049943.1 | - | 7 | 1327 | |||
AK075606.1 | - | 7 | 1345 | |||
AK147686.1 | - | 7 | 1327 | |||
BC010825.1 | - | 7 | 1358 | 4219273 | ||
BC024796.1 | - | 7 | 1334 | 5360438 | ||
BC025023.1 | - | 7 | 1314 | 5364554 | ||
BC054770.1 | - | 13 | 1323 | 6415204 |