Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025797.3 | NP_080073.1 | 4, 5 | 385 | ||
GenBank mRNA | AK002426.1 | - | 4, 5 | 386 | ||
AK013210.1 | - | 4, 5 | 386 | |||
AK132102.1 | - | 4, 5 | 408 | |||
BC024341.1 | - | 4, 5 | 390 | 4973059 |