Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025350.3 | NP_079626.2 | 3 | 462 | ||
GenBank mRNA | AK003088.1 | - | 3 | 247 | ||
BC052661.1 | - | 3 | 476 | 30050094 |