Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025973.3 | NP_080249.2 | 2 | 227 | ||
GenBank mRNA | AK008886.1 | - | 2 | 228 | ||
AK008959.1 | - | 2 | 228 | |||
AK160474.1 | - | 2 | 228 | |||
BC099409.1 | - | 2 | 218 | 30790663 |