Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053248.2 | NP_444478.2 | 7, 8 | 1068 | ||
GenBank mRNA | AF235001.1 | - | 7, 8 | 1035 | ||
AK046718.1 | - | 7, 8 | 1069 |