Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291915.1 | NP_001278844.1 | 3 | 411 | ||
NM_013459.3 | NP_038487.1 | 3 | 414 | |||
GenBank mRNA | AK143817.1 | - | 3 | 358 | ||
BC138778.1 | - | 3 | 414 | 8861800 | ||
BC138779.1 | - | 3 | 414 | 8861801 | ||
BC145412.1 | - | 3 | 411 | 9053970 | ||
DV051476.1 | - | 3 | 359 | |||
M11768.1 | - | 3 | 355 |