Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291915.1 | NP_001278844.1 | 5 | 790 | ||
NM_013459.3 | NP_038487.1 | 5 | 793 | |||
GenBank mRNA | AK143817.1 | - | 5 | 737 | ||
BC138778.1 | - | 5 | 793 | 8861800 | ||
BC138779.1 | - | 5 | 793 | 8861801 | ||
BC145412.1 | - | 5 | 790 | 9053970 | ||
BE136302.1 | - | 1 | 130 | |||
DV051476.1 | - | 5 | 738 | |||
M11768.1 | - | 5 | 734 |