Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005847.2 | NP_001005847.1 | 4 | 586 | ||
NM_001205054.1 | NP_001191983.1 | 4 | 586 | |||
GenBank mRNA | AA396103.1 | - | 4 | 430 | ||
AK213583.1 | - | 2 | 138 | |||
BC048230.1 | - | 4 | 1467 | |||
BC132227.1 | - | 4 | 561 | 40130504 | ||
BC132229.1 | - | 4 | 561 | 40130506 | ||
BC145297.1 | - | 4 | 561 | 9053851 | ||
BQ571087.1 | - | 4 | 478 | |||
S81393.1 | - | 4 | 503 |