Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027548.3 | NP_081824.1 | 8 | 902 | ||
GenBank mRNA | AF105328.1 | - | 7 | 840 | ||
AK014524.1 | - | 8 | 905 | |||
BC064004.1 | - | 8 | 895 | 30288309 |