Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081978.2 | NP_001075447.1 | 5 | 600 | ||
NM_001290371.1 | NP_001277300.1 | 5 | 528 | |||
NM_009666.4 | NP_033796.1 | 5 | 600 | |||
GenBank mRNA | AI323496.1 | - | 2 | 208 | ||
AK029358.1 | - | 5 | 604 | |||
BC059090.1 | - | 5 | 570 | 30286944 | ||
CD773419.1 | - | 5 | 604 | |||
D31768.1 | - | 5 | 591 | |||
D31769.1 | - | 5 | 519 | |||
KC731591.1 | - | 5 | 528 | |||
M10095.1 | - | 2 | 408 |