Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081978.2 | NP_001075447.1 | 4, 5 | 204 | ||
NM_009666.4 | NP_033796.1 | 4, 5 | 204 | |||
GenBank mRNA | AK029358.1 | - | 4, 5 | 208 | ||
BC059090.1 | - | 4, 5 | 174 | 30286944 | ||
CD773419.1 | - | 4, 5 | 208 | |||
D31768.1 | - | 4, 5 | 195 | |||
S74899.1 | - | 5, 6 | 246 |